Learn More
Medchemexpress LLC (1S,2S)-ML-SI3 ((+)-trans-ML-SI3) | 2563870-87-9 | 100.0% | 429.58 g/mol | C23H31N3O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1348195MG
Encompass_Preferred
(1S,2S)-ML-SI3 ((+)-trans-ML-SI3) is a stereoisomeric small-molecule modulator of TRPML ion channels used in biochemical and cellular research. It activates TRPML2 and TRPML3 while inhibiting TRPML1, enabling investigation of lysosomal ion channel function and neuronal signaling. Supplied as a high-purity research reagent in small laboratory pack sizes.
  • High purity suitable for biochemical and cellular assays.
  • Selective modulation of TRPML2 and TRPML3, with inhibitory activity at TRPML1.
  • Stereochemically defined isomer for reproducible results.
  • Good solubility in DMSO for in vitro preparation.
  • Available in small lab-scale quantities for research use.
Catalog No. 50-002-23399