Learn More
Medchemexpress LLC (1S,3R,5R)-PIM447 (dihydrochloride) | 513.38 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY19322C5MG
Encompass_Preferred
PIM447 (LGH447) is a potent, orally available, and selective pan-PIM kinase inhibitor. It displays dual antimyeloma and bone-protective effects and induces apoptosis.
  • Potent, orally available, and selective pan-PIM kinase inhibitor.
  • Ki values of 6, 18, and 9 pM for PIM1, PIM2, and PIM3, respectively.
  • Displays dual antimyeloma and bone-protective effects.
  • Induces apoptosis.
Catalog No. 50-003-85442