Learn More
Cayman Chemical 1Stearoyl215 SHETEsngl 100ug
SDP
Supplier:  Cayman Chemical 21139100 UG
Encompass_Preferred
An oxidized phospholipid; formed in A23187-activated monocytes by direct oxidation of endogenous PE
Catalog No. 50-298-3335