Learn More
Cayman Chemical 1Stearoyl215 SHETEsngl 50ug
SDP
Supplier:  Cayman Chemical 2793150 UG
Encompass_Preferred
An internal standard for the quantification of 1-stearoyl-2-15(S)-HETE-sn-glycero-3-PE by GC- or LC-MS
Catalog No. 50-293-9194