Learn More
Cayman Chemical 1Stearoyl215 SHpETEsng 100ug
SDP
Supplier:  Cayman Chemical 25856100 UG
Encompass_Preferred
A phospholipid that contains stearic acid at the sn-1 position and 15(S)-HpETE at the sn-2 position; is produced via oxidation of SAPE by 15-LO; increases ferroptotic cell death in RSL3-treated wild-type and Acls4 knockout Pfa1 MEFs at 0.6 and 0.9 μM
Catalog No. 50-293-3445