Learn More
Cayman Chemical 1Stearoyl2Arachidonoyld8sngly
SDP
Supplier:  Cayman Chemical 10009872500 UG
Encompass_Preferred
An internal standard for the quantification of SAG by GC- or LC-MS
Catalog No. 50-288-7897