Learn More
Cayman Chemical 1Stearoyl2ArachIdonylsn 10mg
SDP
Supplier:  Cayman Chemical 2587110 MG
Encompass_Preferred
A naturally-occurring phospholipid; formed via the phosphatidylserine decarboxylation pathway in mammalian cells; has been used in the generation of giant unilamellar vesicles
Catalog No. 50-293-4335