Learn More
Cayman Chemical 1Stearoyl2Oleoyl3LInoleo 10mg
SDP
Supplier:  Cayman Chemical 2696810 MG
Encompass_Preferred
A triacylglycerol; has been found in peanut and soybean oils as well as ostrich and emu oils; has been used as a substrate for soybean lipoxygenase in cell-free assays
Catalog No. 50-294-1729