Learn More
Cayman Chemical 1Stearoyl2PalmItoyl3Oleo 5mg
SDP
Supplier:  Cayman Chemical 272905 MG
Encompass_Preferred
A triacylglycerol that contains stearic acid at the sn-1 position, palmitic acid at the sn-2 position, and oleic acid at the sn-3 position; has been found in bovine milk
Catalog No. 50-293-9604