Learn More
Cayman Chemical 1T-LSdolutIon 100ug
SDP
Supplier:  Cayman Chemical 40712100 UG
Encompass_Preferred
An analytical reference standard categorized as a lysergamide; has been found in paper sheets obtained by a forensic lab in Japan; intended for research and forensic applications
Catalog No. 50-294-4963