Learn More
Assay Biotech Trk C (Phospho-Tyr516) Antibody, 50 uL
Supplier:  Assay Biotech A8196
Encompass_Preferred
Anti-Trk C (Phospho-Tyr516) polyclonal Antibody detects Trk C only when phosphorylated at Tyr516. Validated for WB, IHC and ELISA. Reactive for Human, Mouse, Rat.
Catalog No. NC1826351