Learn More
Chemglass Life Sciences Chemglass 2.0 mL Cryogenic Vial - Self-Standing, Internal Thread, Sterile (Racked, 1,200/Case)
SDP
Supplier:  Chemglass Life Sciences CGN4766008
Encompass_Preferred
2.0 mL sterile cryogenic vial with an internal thread and self-standing round bottom, supplied in a racked format for convenient handling and ultra-low temperature storage.
  • Self-standing round bottom design for stable upright storage.
  • Internal thread closure for secure sealing.
  • Sterilized by E-beam; non-pyrogenic and DNase/RNase free.
  • Clear graduations and a white marking area for sample identification.
  • Solid, one-piece construction to prevent barcode loss at low temperatures.
  • Autoclavable for flexible sterilization workflows.
  • Racked packaging: 10 x 10 vials per rack (100 vials/rack), 12 racks per case (1,200 vials/case).
  • Recommended for storage in the gas phase of liquid nitrogen.
Catalog No. 50-273-3345