Learn More
Chemglass Life Sciences 2.0 mL Self-Standing Sterile Vials, Natural Caps, External Thread, 50/pk
SDP
Supplier:  Chemglass Life Sciences CGN4764002N
Encompass_Preferred
2.0 mL self-standing sterile sample vials with natural-color caps and external thread. The sealing ring provides improved leak resistance for routine sample storage and handling. Packaged for laboratory use in multiple quantities for convenience.
  • Self-standing design for stable upright storage.
  • External thread with sealing ring for improved leak resistance.
  • Natural-color caps for easy sample identification.
  • Sterile packaging suitable for sample handling.
  • Available packaging: 50 per pack, 500 per box, 2,000 per case.
Catalog No. 50-273-3361