Learn More
Chemglass Life Sciences Chemglass 2.0 mL Self-Standing Sample Vials, External Thread, Hinged Cap with Sealing Ring, Natural Caps, Sterile, 50/pk
SDP
Supplier:  Chemglass Life Sciences CGN4764012N
Encompass_Preferred
Chemglass 2.0 mL self-standing sample vials (CGN-4764-012N) are sterile, external-thread vials with a hinged cap and sealing ring supplied with natural color caps and nest packaging for clean handling and laboratory storage.
  • 2.0 mL capacity.
  • External thread with hinged cap and sealing ring.
  • Natural color caps; photo color may vary.
  • Sterile; electron beam sterilized, DNAse-free, RNAse-free, pyrogen-free, and endotoxin-free.
  • Packaged 50 per pack, 500 per box, 2000 per case.
  • Self-standing round bottom design in nest packaging for sterile handling.
Catalog No. 50-273-3357