Learn More
Medchemexpress LLC 2-[(1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-4-methyl-1H-pyrrole-3-propanoic acid | 215543-92-3 | MFCD08235144 | >95.0% | 296.3 g/mol | C17H16N2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10407100MG
Encompass_Preferred
SU 5402 is a small-molecule receptor tyrosine kinase inhibitor used in research to probe VEGF, FGF, and PDGF signaling pathways. It inhibits VEGFR2, FGFR1, and PDGFRβ with reported IC50 values of ~20 nM, 30 nM, and 510 nM, respectively, and is supplied as a research reagent for in vitro studies.
  • Potent inhibition of VEGFR2, FGFR1, and PDGFRβ (IC50 ≈ 20 nM, 30 nM, 510 nM).
  • Useful for studying angiogenesis and receptor tyrosine kinase signaling.
  • Cell-permeable small molecule suitable for in vitro assays.
  • Available as a solid for solution preparation and dosing flexibility.
  • Supplied with documentation; verify purity and handling per SDS before use.
Catalog No. 50-002-42595