Learn More
MSC PRO-SOURCE 05106 2-1/2" Long x 3/4" Diam Stainless Steel Twisted Wire Bristle Brush Single Spiral, 9" Overall Length, 0.008" Wire Diam, 0.142" Shank Diam
Supplier:  MSC 03716081
Encompass
PRO-SOURCE 05106 2-1/2" Long x 3/4" Diam Stainless Steel Twisted Wire Bristle Brush Single Spiral, 9" Overall Length, 0.008" Wire Diam, 0.142" Shank Diam
Catalog No. NC1731407