Learn More
Medchemexpress LLC 2-({[1-(4-fluorophenyl)-5-(2-methoxyphenyl)-1H-pyrazol-3-yl]carbonyl}amino)tricyclodecane-2-carboxylic acid | 1811503-67-9 | MFCD11042212 | 95.6% | 489.54 | C28H28FN3O4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1248315MG
Encompass_Preferred
NTRC-844 is a small-molecule research compound that functions as a selective antagonist of neurotensin receptor type 2 (NTS2). It is employed in preclinical analgesic and neuropharmacology studies and has a reported EC50 of 238 nM. Chemical identity: C28H28FN3O4; molecular weight 489.54 g/mol. Purity is batch dependent (manufacturer COA example: 95.6%). Supplied in milligram research quantities for laboratory use.
  • Selective antagonist of neurotensin receptor type 2 (EC50 238 nM).
  • Intended for preclinical analgesic and neuropharmacology research.
  • Chemical formula C28H28FN3O4 and molecular weight 489.54 g/mol.
  • Provided with a batch-specific certificate of analysis documenting purity.
  • Available in milligram-scale research pack sizes.
Catalog No. 50-002-37112