Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-benzofuran-2-yl)-4,5-dihydro-1H-imidazole hydrochloride | 89196-95-2 | MFCD00672666 | >=98.0% | 222.67 g/mol | C11H11ClN2O | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1009045MG

Encompass_Preferred

RX 801077 hydrochloride (2-BFI) is a research-grade imidazoline I2 receptor (I2R) agonist used in preclinical and in vitro studies to investigate neuroprotective and anti-inflammatory effects. The hydrochloride salt is supplied as a solid and as solution formulations for laboratory use.

  • Selective imidazoline I2 receptor agonist with reported Ki ≈ 70.1 nM.
  • Demonstrates anti-inflammatory and neuroprotective activity in preclinical studies.
  • Available as solid and as 10 mM solution in DMSO.
  • Supplied in multiple laboratory pack sizes, including 5 mg.
  • Molecular weight 222.67 g/mol.
  • CAS number 89196-95-2.
  • Purity ≥98% by HPLC.

Catalog No. 50-002-13003


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.