Learn More
Medchemexpress LLC Α2β1 integrin ligand peptide TFA (DGEA peptide TFA) | 98.6% | 504.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1868A5MG
Encompass_Preferred
α2β1 Integrin Ligand Peptide TFA interacts with the α2β1 integrin receptor on the cell membrane, mediating extracellular signals into cells. It is a potential antagonist of collagen receptors. In vitro studies demonstrate its ability to inhibit various viral infections and induce tyrosine kinase-dependent calcium mobilization in specific cell types.
  • Interacts with the α2β1 integrin receptor
  • Mediates extracellular signals into cells
  • Potential antagonist of collagen receptors
  • Inhibits SA11, RRV, and Wa infection in a dose-dependent manner
  • Induces tyrosine kinase-dependent calcium mobilization in osteoblasts and fibroblasts
Catalog No. 50-004-23984