Learn More
Medchemexpress LLC 2-(1-methylethyl)phenyl diphenyl phosphate | 64532-94-1 | MFCD00870203 | >98.0% | 368.36 | C21H21O4P | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW588267100MG
Encompass_Preferred
2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant and plasticizer supplied for research use; consult the product datasheet and Certificate of Analysis for handling, purity, and storage details.
  • Organophosphate flame retardant and plasticizer.
  • Used in materials testing and formulation research to impart flame retardancy.
  • CAS number 64532-94-1; molecular formula C21H21O4P; molecular weight 368.36.
  • Supplied in small-quantity research packs suitable for analytical work.
  • For research use only; consult SDS and CoA for purity and storage information.
Catalog No. 50-002-84029