Learn More
Medchemexpress LLC 2-(1-Piperazinyl)pyrimidine | 20980-22-7 | 164.21 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00446410MM/1ML
Encompass_Preferred
2-(1-Piperazinyl)pyrimidine is a chemical compound that serves as the major metabolite of Tandospirone. This endogenous metabolite is utilized in various research applications.
  • Functions as the major metabolite of Tandospirone.
  • Identified as an endogenous metabolite.
  • Applicable for in vitro studies, with solubility in DMSO.
  • Applicable for in vivo studies, with specific dissolution protocols.
Catalog No. 50-004-48826