Learn More
Cayman Chemical 2 1 ThIophen2ylethylIden 10mg
SDP
Supplier:  Cayman Chemical 3138510 MG
Encompass_Preferred
An antimicrobial agent; active against Gram-negative bacteria (MICs = 64-100 μg/ml), Gram-positive bacteria (MICs = 50-130 μg/ml), and fungi (MICs = 69-120 μg/ml); a precursor in the synthesis of other antimicrobial agents, as well as compounds with anticancer activity
Catalog No. 50-299-1893