Learn More
Medchemexpress LLC 2-(11-oxo-6H-benzo[c]benzoxepin-2-yl)acetic acid | 55453-87-7 | MFCD00242952 | 268.27 g/mol | C16H12O4 | 1 G

Supplier: Medchemexpress LLC HYW0500881G
Isoxepac is an orally active non-steroidal anti-inflammatory research standard supplied as a powder for preclinical and analytical use. It is used in studies of inflammation and pain-related disease models.
- cas number: 55453-87-7
- chemical formula: C16H12O4
- molecular weight: 268.27 g/mol
- form: powder
- storage: powder at -20°C for long-term, 4°C short-term; in solvent at -80°C or -20°C per guidance
- solubility: soluble in DMSO (100 mg/mL); formulations available for in vivo dosing
- applications: inflammatory and pain-related preclinical research and analytical assays
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.