Learn More
Medchemexpress LLC 2-(1H-benzimidazol-2-ylthio)-N'-[(5-bromo-2-hydroxyphenyl)methylidene]propanehydrazide | 330676-02-3 | 99.0% | 419.30 g·mol⁻¹ | C17H15BrN4O2S | 50 MG

Supplier: Medchemexpress LLC HY10319450MG
KH7 is a soluble adenylyl cyclase (sAC) inhibitor used as a research tool to modulate intracellular cAMP signaling in biochemical and cellular assays.
- Selective soluble adenylyl cyclase (sAC) inhibitor with IC50 3-10 μM in biochemical and cellular assays.
- Useful for studying cAMP-dependent signaling and related cellular processes.
- Reported purity 99.0% (typical, HPLC).
- Solid powder, commonly supplied in 50 mg packages.
- Molecular formula C17H15BrN4O2S; molecular weight 419.30 g·mol⁻¹.
- CAS number 330676-02-3 for substance identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.