Learn More
Medchemexpress LLC 2(1H)-Pyrimidinone, 4-[2-(3-bromophenyl)ethenyl]-6-(trifluoromethyl)- | 357158-20-4 | C13H8BrF3N2O | 10 MM / 1 ML
SDP
Supplier:  Medchemexpress LLC HY15368610MM/1ML
Encompass_Preferred
BVFP binds to the PGRN588-593 peptide with a Kd of 20 μM. It can disrupt PGRN-SORT1 binding and inhibits SORT1-mediated rPGRN endocytosis.
  • Binds to the PGRN588-593 peptide with a Kd of 20 μM.
  • Disrupts PGRN-SORT1 binding.
  • Inhibits SORT1-mediated rPGRN endocytosis.
Catalog No. 50-003-76019