Learn More
Medchemexpress LLC 2(1H)-Quinolinone, 7-amino-4-methyl- | 19840-99-4 | 174.20 | 1 G

Supplier: Medchemexpress LLC HYW0082921G
Carbostyril 124 is a sensitizing chromophore that acts as a reasonably effective organic sensitizer for a proximate bound terbium ion. This compound is intended for research use only.
- Yellow to brown solid appearance
- Store at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
- Soluble in DMSO (50 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.