Learn More
Medchemexpress LLC 2(1H)-Quinolinone, 7-amino-4-methyl- | 19840-99-4 | 174.20 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0082925G
Encompass_Preferred
Carbostyril 124 is a sensitizing chromophore that functions as a reasonably effective organic sensitizer for a proximate bound terbium ion. It exhibits characteristic metal-based luminescence for europium and terbium complexes in aqueous solution when excited at 320 nm.
  • Sensitizing chromophore
  • Effective organic sensitizer for a proximate bound terbium ion
  • Exhibits metal-based luminescence with europium and terbium complexes
Catalog No. 50-004-46871