Learn More
Medchemexpress LLC 2(1H)-Quinolinone, 7-amino-4-methyl- | 19840-99-4 | 174.20 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW008292500MG
Encompass_Preferred
Carbostyril 124 is a sensitising chromophore used for research purposes only. It acts as an effective organic sensitizer for a proximate bound terbium ion and shows characteristic metal-based luminescence for europium and terbium complexes.
  • Sensitising chromophore.
  • Effective organic sensitizer for a proximate bound terbium ion.
  • Exhibits metal-based luminescence for europium and terbium complexes.
  • Soluble in DMSO at 50 mg/mL.
  • For research use only.
Catalog No. 50-004-49399