Learn More
Medchemexpress LLC 2(1H)-Quinolinone, 8-hydroxy- | 15450-76-7 | 99.9% | 161.16 | 10 G
Supplier: Medchemexpress LLC HY3449410G
2,8-Quinolinediol, also known as 8-Hydroxy-2(1H)-quinolinone, is a biochemical reagent. It is utilized as a biological material or organic compound for diverse life science research applications.
- Used as a biochemical reagent.
- Suitable for life science related research.
- Available as a solid, white to off-white powder.
- Stable for 3 years at -20°C and 2 years at 4°C in powder form.
- Stable for 6 months at -80°C and 1 month at -20°C when in solvent.
- Highly soluble in DMSO.