Learn More
Medchemexpress LLC 2(1H)-quinolinone, 8-hydroxy-5-[(1R,2S)-1-hydroxy-2-[(1-methylethyl)amino]butyl] hydrochloride | 62929-91-3 | 100.0% | 326.82 | C16H23ClN2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1007045MG
Encompass_Preferred
Procaterol hydrochloride is a selective, intermediate-acting β2-adrenoceptor agonist used in research for bronchodilation and modulation of inflammatory responses. It shows an EC50 of 12.9 μM at β2 receptors and has demonstrated anti-inflammatory effects in vitro and in vivo. High-purity material is provided for biochemical and cellular studies.
  • Selective intermediate-acting β2-adrenoceptor agonist.
  • Demonstrated anti-inflammatory activity in vitro and in vivo.
  • Reported EC50 of 12.9 μM for β2-adrenoceptor activity.
  • High purity suitable for research and assay use.
  • Available in small, weighed quantities for experimental flexibility.
Catalog No. 50-002-14038