Learn More
Medchemexpress LLC 2-[(1R)-3-oxo-1-phenyl-3-[4-(trifluoromethyl)phenyl]propyl]propanedioic acid | 1410101-89-1 | 98.1% | C19H15F3O5 | 10MG

Supplier: Medchemexpress LLC HY1385610MG
(R)-PS210 is the R enantiomer of PS210, a substrate-selective allosteric activator of PDK1 used for biochemical and cell-signaling research targeting the PI3K/Akt/mTOR pathway. Supplied as a white to off-white solid with reported high purity, it is suitable for in vitro assays and comes in pre-weighed and solution formats for convenient use.
- Activates PDK1 with AC50 1.8 μM.
- High purity suitable for research: 98.1%.
- Molecular formula C19H15F3O5; molecular weight 380.31 g·mol⁻¹.
- Supplied as a white to off-white solid for handling convenience.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Available as a ready-to-use 10 mM solution in DMSO for assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.