Learn More
Medchemexpress LLC 2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethanol | 86770-74-3 | MFCD01752874 | >98.0% | 193.24 g/mol | C8H19NO4 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW00800525G
Encompass_Preferred
Amino-PEG4-alcohol is a tetraethylene glycol monoamine with a terminal primary amine and a terminal hydroxyl. It is used as a PEG-based linker and functional handle in PROTAC synthesis, bioconjugation, and other chemical syntheses requiring an amine and alcohol functionality.
  • Provides terminal amino and alcohol functional groups.
  • Suitable for PROTAC linker and bioconjugation applications.
  • Molecular weight 193.24 g/mol and formula C8H19NO4.
  • Typical purity greater than 98.0% (GC).
  • Available in laboratory pack sizes including 25 G.
Catalog No. 50-002-02841