Learn More
Medchemexpress LLC 2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethanol | 86770-74-3 | MFCD01752874 | 99.0% | 193.24 g/mol | C8H19NO4
SDP
Supplier:  Medchemexpress LLC HYW008005500MG
Encompass_Preferred
Amino-PEG4-alcohol (2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethanol) is a tetraethylene glycol derivative with a terminal primary amine and a terminal hydroxyl group. It is used as a PEG4 spacer/linker in PROTAC and antibody-drug conjugate (ADC) synthesis.
  • Provides a terminal primary amine and a terminal alcohol for bioconjugation.
  • Four ethylene glycol units give a flexible, hydrophilic spacer.
  • Used in PROTAC and ADC linker design.
  • Typical molecular formula C8H19NO4 and molecular weight 193.24 g/mol.
  • Available from multiple suppliers in solid and solution formats; pack sizes vary by vendor.
  • Reported purity by suppliers typically ranges from >98% (HPLC) to 99% depending on source.
Catalog No. 50-002-03136