Learn More
Medchemexpress LLC 2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethanol | 86770-74-3 | MFCD01752874 | 99.8% | 193.24 g/mol | C8H19NO4 | 1 G

Supplier: Medchemexpress LLC HYW0080051G
Amino-PEG4-alcohol is a tetraethylene glycol monoamine (H2N-PEG4-OH) with a terminal primary amine and a terminal hydroxyl group. It is used as a 4-unit PEG linker for bioconjugation, in PROTAC synthesis, and as a non-cleavable PEG ADC linker.
- Peg4 linker with a terminal primary amine and a terminal alcohol.
- Molecular weight 193.24 g/mol.
- Chemical formula C8H19NO4.
- CAS number 86770-74-3.
- High purity (≈99.8%), suitable for conjugation reactions.
- Available in multiple pack sizes, including 1 G for small-scale use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.