Learn More
Medchemexpress LLC 2-[2-[2-(5-bromo-2-methoxyphenyl)ethyl]-3-fluorophenyl]-4,5-dihydro-1H-imidazole hydrochloride | 1706524-94-8 | MFCD28133377 | 99.3% | 413.71 g·mol⁻¹ | C18H19BrClFN2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1101235MG
Encompass_Preferred
ML-00253764 hydrochloride is the hydrochloride salt of a brain-penetrant, nonpeptidic antagonist of the melanocortin 4 receptor (MC4R) used in pharmacological research. It exhibits submicromolar binding and functional inhibition of MC4R, is supplied as a high-purity solid, and is intended for in vitro and in vivo studies when handled and stored according to manufacturer recommendations.
  • Brain-penetrant, nonpeptidic melanocortin 4 receptor (MC4R) antagonist.
  • Submicromolar potency: Ki = 0.16 μM, IC50 = 0.103 μM.
  • High purity (~99.3%) suitable for research applications.
  • Hydrochloride salt form for improved handling and solubility.
  • Solid, off-white to light yellow appearance; easy to weigh and formulate.
  • Recommended for in vitro and in vivo pharmacology with proper storage.
Catalog No. 50-002-63559