Learn More
Medchemexpress LLC 2,2,2-Trifluoro-1-(4'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl)ethanone | 893407-18-6 | 97.7% | C15H11F3O3S | 5 G
SDP
Supplier:  Medchemexpress LLC HY770245G
Encompass_Preferred
2,2,2-Trifluoro-1-(4'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl)ethanone is a drug intermediate used for the synthesis of various active compounds.
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years
  • Stable in solvent at -20°C for 1 year
  • Can be shipped at room temperature if duration is less than 2 weeks
Catalog No. 50-004-33461