Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[2-(4-aminophenyl)-3H-benzimidazol-5-yl]-3H-benzimidazol-5-amine | 293762-45-5 | 99.1% | 340.38 g/mol | C20H16N6 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY10324125MG

Encompass_Preferred

Ro 90-7501 is a small-molecule research compound that inhibits amyloid β42 (Aβ42) fibril assembly and reduces Aβ42-induced cytotoxicity. It has reported activity affecting DNA damage response and phosphatase regulation, and has been used in studies of radiosensitization and antiviral innate immune modulation.

  • Inhibits amyloid β42 fibril assembly and reduces Aβ42-induced cytotoxicity (EC50 ≈ 2 μM).
  • Reported to inhibit ATM phosphorylation and impair DNA repair.
  • Modulates innate immune signaling and inhibits protein phosphatase 5 (PP5) in a TPR-dependent manner.
  • Used in neurodegeneration, amyloid aggregation, and radiosensitization research applications.
  • Molecular formula C20H16N6 and molecular weight 340.38 g/mol.
  • Purity typically reported ≈99.1% and soluble in DMSO.

Catalog No. 50-002-46299


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.