Learn More
Medchemexpress LLC 2-[2-(4-iodoanilino)-2-oxoethoxy]-N-[2-(N-methylanilino)-2-oxoethyl]benzamide | 1462267-07-7 | 99.5% | 10 MG

Supplier: Medchemexpress LLC HY12003410MG
NCGC 607 is a noninhibitory small-molecule chaperone of glucocerebrosidase (GCase) used in research to increase GCase activity, reduce α-synuclein, and decrease glycolipid levels. It is supplied as a high-purity white to off-white solid for research use.
- Noninhibitory glucocerebrosidase (GCase) chaperone
- Increases GCase activity and reduces α-synuclein levels
- High purity solid suitable for in vitro research
- Soluble in DMSO (≈62.5 mg/mL) with ultrasonic assistance
- Store at 4°C and protect from light; in solvent store at -80°C for long-term
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.