Learn More
Medchemexpress LLC 2-[2-(4-methoxy-2-pyridinyl)ethyl]-1H-imidazo[4,5-b]pyridine;dihydrochloride | 1216722-25-6 | MFCD11100187 | 99.6% | 327.21 g/mol | C14H16Cl2N4O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10758410MG
Encompass_Preferred
BYK 191023 dihydrochloride is a small-molecule inducible nitric-oxide synthase (iNOS) inhibitor supplied as the dihydrochloride salt for biochemical and pharmacological research. It is characterized by CAS 1216722-25-6, molecular formula C14H16Cl2N4O, and a molecular weight of 327.21 g/mol, and is offered as a high-purity reagent suitable for in vitro studies.
  • High purity (≈99.6% by HPLC).
  • Dihydrochloride salt for improved solubility and handling.
  • Characterized molecular identity (CAS 1216722-25-6; C14H16Cl2N4O).
  • Suitable for in vitro studies of nitric-oxide synthase pathways.
  • Stable under recommended storage; follow supplier storage instructions.
Catalog No. 50-002-40026