Learn More
Medchemexpress LLC 2,2',4'-Trichloroacetophenone | 4252-78-2 | 99.8% | 223.48 | 100 G
SDP
Supplier:  Medchemexpress LLC HY33040100G
Encompass_Preferred
2,2',4'-Trichloroacetophenone is an α-haloacetophenone analogue that demonstrates effective antibacterial activities. It is particularly active against Xanthomonas oryzae pv. oryzae (Xoo) and Xanthomonas axonopodis pv. citri (Xac), with EC50 values of 0.54 and 2.02 mg/L, respectively. This product is intended for antibacteria study.
  • Exhibits antibacterial activities.
  • Effective against specific plant pathogenic bacteria.
  • Suitable for research purposes.
Catalog No. 50-004-04723