Learn More
Strem, An Ascensus Company CAS# 1148-79-4. 1g. 2,2':6',2''-Terpyridine, min. 98% TERPY. MFCD00006213
SDP
Supplier:  Strem, An Ascensus Company 0719201G
Encompass
CAS# 1148-79-4. 1g. 2,2':6',2''-Terpyridine, min. 98% TERPY. MFCD00006213. Molecular Weight: 233.27. Molecular Formula: C5H4NC5H3NC5H4N. Color/form: off-white pwdr. Strem# 07-1920. http://www.strem.com/catalog/v/07-1920/
Catalog No. 50-901-09085