Learn More
Medchemexpress LLC 2-(2-((6-Chlorohexyl)oxy)ethoxy)acetic acid | 2231744-57-1 | 95.0% | 238.71 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4304850MG
Encompass_Preferred
2-(2-((6-Chlorohexyl)oxy)ethoxy)acetic acid is a PROTAC linker utilized in PROTAC SMARCA2/4-degrader-34 (HY-169279). It is intended for research use only and is not for patient use. This compound has been referenced in studies exploring small molecule degraders to enhance chemotherapy efficacy.
  • PROTAC linker
  • For research use only
  • Not sold to patients
  • Referenced in studies on chemotherapy efficacy
Catalog No. 50-004-03896