Learn More
Medchemexpress LLC 2-[[(2,6-dichlorophenyl)methyl]thio]-5-(1H-pyrrol-2-yl)-1,3,4-oxadiazole | 2253744-54-4 | 99.9% | 326.20 | C13H9Cl2N3OS | 50MG
SDP
Supplier:  Medchemexpress LLC HY12601050MG
Encompass_Preferred
Dooku1 is a small-molecule reversible antagonist of Yoda1 that inhibits Yoda1-induced Ca2+ entry in cellular assays. It is provided for research use and is characterized by molecular formula C13H9Cl2N3OS, molecular weight 326.20, and high chemical purity for reliable in vitro experiments.
  • Reversible antagonist of Yoda1, suitable for mechanistic studies.
  • Demonstrated cellular activity with reported IC50 approximately 1.3 μM for Yoda1-induced Ca2+ entry.
  • High purity (>99.9%) for reproducible results.
  • Supplied as a compact 50 mg pack for small-scale experiments.
  • Applicable to in vitro studies of mechanosensitive ion channel signaling.
Catalog No. 50-002-38004