Learn More
Medchemexpress LLC 2-[(2,6-dimethylphenyl)amino]-N,N,N-trimethyl-2-oxoethaniminium chloride | 5369-00-6 | 96.3% | 256.77 g/mol | C13H21ClN2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10136210MM 1ML
Encompass_Preferred
QX-222 chloride is a quaternary trimethyl analogue of lignocaine that functions as a potent voltage-gated sodium (Na+) channel blocker used in electrophysiology and ion-channel research. It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as a solid for formulation and dosing flexibility.
  • Potent Na+ channel blocker for electrophysiology studies.
  • Quaternary trimethyl analogue of lignocaine with intracellular blocking properties.
  • Provided as 10 mM solution in DMSO (1 mL) for ready-to-use stock preparations.
  • Also available as solid material for custom formulations.
  • High purity suitable for research use.
  • Stable when stored under recommended conditions to preserve activity.
Catalog No. 50-002-11991