Learn More
Medchemexpress LLC 2,2'-Anhydrouridine | 3736-77-4 | 99.5% | 226.19 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01231310MM/1ML
Encompass_Preferred
2,2'-Anhydrouridine (2,2'-Cyclouridine) is a nucleoside analog and an important intermediate in the synthesis of other nucleoside-related compounds. The 5-substituted analogues of 2,2'-Anhydrouridine are competitive inhibitors of uridine phosphorylase.
  • Nucleoside analog.
  • Important intermediate for synthesizing other nucleoside-related compounds.
  • 5-substituted analogues are competitive inhibitors of uridine phosphorylase.
Catalog No. 50-004-52365