Learn More
Medchemexpress LLC 2-(2-azidoethoxy)ethyl 4-methylbenzenesulfonate | 182347-24-6 | MFCD27946436 | 99.8% | 285.32 g·mol⁻¹ | C11H15N3O4S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1908361G
Encompass_Preferred
N3-PEG2-Tos is an azide-terminated PEG2 linker bearing a tosylate leaving group. It is designed as a small, bifunctional spacer for organic synthesis and bioconjugation: the azide provides a click-chemistry handle for azide-alkyne cycloadditions, while the tosylate permits nucleophilic substitution to introduce other functional groups.
  • Azide functional group enables click chemistry conjugation.
  • Tosylate leaving group allows facile nucleophilic substitution.
  • Short PEG2 spacer provides flexibility and solubility.
  • High purity suitable for synthetic and assay applications.
  • Available as a 1 g solid for small-scale synthesis.
  • Molecular weight ~285.3 g·mol⁻¹ for stoichiometry calculations.
Catalog No. 50-002-83072