Learn More
Medchemexpress LLC 2-(2-azidoethoxy)ethyl 4-methylbenzenesulfonate | 182347-24-6 | MFCD27946436 | 99.8% | 285.32 g/mol | C11H15N3O4S | 100 MG

Supplier: Medchemexpress LLC HYW190836100MG
N3-PEG2-Tos (2-(2-azidoethoxy)ethyl 4-methylbenzenesulfonate) is a small polyethylene glycol (PEG) linker bearing an azide functional group and a tosylate leaving group. It is used as a bifunctional building block for introducing an azide handle and for electrophilic substitution during linker and conjugation chemistry. CAS 182347-24-6; molecular weight 285.32 g/mol.
- Provides an azide handle for click chemistry applications.
- Contains a tosylate leaving group for efficient nucleophilic substitution.
- Small PEG spacer improves solubility and linker flexibility.
- Suitable for linker synthesis and bioconjugation workflows.
- Store at -20 °C, protected from light and moisture.
- High purity reported (~99.8%) for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.