Learn More
Medchemexpress LLC 2,2'-Azobis(2-methylpropionamidine) dihydrochloride | 2997-92-4 | MFCD00142725 | 98.0% | 271.19 g/mol | C8H20Cl2N6 | 1g
SDP
Supplier:  Medchemexpress LLC HYY05251G
Encompass_Preferred
AAPH (2 2 -Azodiisobutyramidine dihydrochloride) has an effect of radical generation AAPH induces oxidative stress and erythrocyte hemolysis[1] AAPH decomposes at 37 to generate an alkyl radical is used as an initiator In the presence of oxygen these alkyl radicals will be converted to peroxyl radicals that can cause lipid peroxidation and loss of erythrocyte membrane integrity which could ultimately lead to hemolysis[1][2][3][4]
Catalog No. 50-002-69063