Learn More
Medchemexpress LLC 2-(2-(((Benzyloxy)carbonyl)amino)acetamido)acetic acid | 2566-19-0 | 99.9% | 266.25 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008731
Encompass_Preferred
2-(2-(((Benzyloxy)carbonyl)amino)acetamido)acetic acid is a derivative of Glycine. Amino acids and their derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
  • Derivative of Glycine
  • Used as ergogenic supplements
  • Influences anabolic hormone secretion
  • Influences fuel supply during exercise
  • Enhances mental performance during stress
  • Prevents exercise-induced muscle damage
  • For research use only
  • Amino acid derivative
  • Reagent for peptide synthesis
  • Peptide fragment
Catalog No. 50-004-51365