Learn More
Medchemexpress LLC 2-(2-(benzyloxy)ethoxy)ethyl 4-methylbenzenesulfonate | 98627-22-6 | MFCD28064331 | 98.5% | 350.43 g/mol | C18H22O5S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0438411G
Encompass_Preferred
Benzyl-PEG2-Tos is a benzyl-protected PEG2-derived tosylate used as a linker and synthetic building block in medicinal chemistry. It provides a short PEG spacer with a tosyl leaving group that facilitates nucleophilic substitution for assembly of PROTACs and bioconjugates.
  • Short PEG2 spacer with benzyl protection.
  • Tosylate leaving group for nucleophilic substitution.
  • Suitable for PROTAC synthesis and linker assembly.
  • High reported purity for research use.
  • Available in gram-scale pack sizes for laboratory research.
Catalog No. 50-002-10169